Benchtop turbidimeters, TU52
Toimittaja: Hach
LZY898 LZY835 LZY901 LZY837 LZY899 LZY877
HACHLZY898EA 940 EUR
HACHLZY898 HACHLZY835 HACHLZY901 HACHLZY837 HACHLZY899 HACHLZY877
Benchtop turbidimeters, TU52
Sameusmittarit
These laser turbidimeters feature performant optical technology. Radio-frequency identification (RFID) uses electromagnetic fields to identify and track tags.
  • Outstanding 90×360° detection technology
  • Matching results between the laboratory and process instruments
  • Data management through USB or Ethernet connections
They can be used for laboratory and process measurements in low turbidity drinking water applications.
Sertifikaatit: ISO 7027 and EPA 180.1
Toimitustiedot: Supplied with a user manual, a Stablcal® set (10 ; 20 ; 600 NTU), power supply,More Product Information