Test kits, photometric, phosphate, LCK cuvette tests
Toimittaja: Hach
LCK348 LCK349 LCK350 LCK351 LCK549
705-3059EA 163 EUR
705-3059 705-3060 705-3061 HACHLCK351 HACHLCK549
Test kits, photometric, phosphate, LCK cuvette tests
Ympäristöanalytiikan pikatestit
Cuvette tests for water testing applications.
  • Precise and reliable measurement values
  • Maximum safety for users, thanks to the closed system and low amounts of reagents
  • Convenient and error-free dosing of the reagents
  • Barcode label for automatic recognition in the photometer
  • Differentiation between tests and measuring ranges by means of colour-coding
Shelf life: 24 months from production date; 36 months for LCK049.
Storage conditions: 15 to 25 °C
Sertifikaatit: ISO 6878-1-1986, DIN 38405 D11-4
Lisävarusteiden tiedot: Cuvette tests are officially approved for many legally required consent limits, with the help of Addista standard solutions and round-robin test solutions, they provide the assurance you need.