Ioniselektiivinen elektrodi (ISE), kloridi, IntelliCAL™
Toimittaja: Hach
ISECL18101 ISECL18103 ISECL181AP
HACHISECL18101EA 1440 EUR
HACHISECL18101 HACHISECL18103 HACHISECL181AP
Ioniselektiivinen elektrodi (ISE), kloridi, IntelliCAL™
Elektrodit ja niiden oheistarvikkeet Ioniselektiiviset elektrodit, ISE
IntelliCAL™ ISECL181 is a digital, combination chloride ion selective electrode (ISE) with a non refillable, double junction reference and built-in temperature sensor. The solid-state sensor design eliminates membrane replacement and allows for dry storage of the ISE.
  • Compatible with digital Hach meters only (e.g. HQd meters)
  • Requires virtually no maintenance
  • Fast, stable, and accurate results
  • Ultimate traceability in measurement history
  • Can be moved between meters without the need to re-calibrate or re-enter measurement settings
The probe isMore Product Information