Ioniselektiivinen elektrodi (ISE), referenssi elektrodit, Radiometer Analytical
Toimittaja: Hach
E21M009 E21M001 E21M002 B20B300
HACHE21M009EA 340 EUR
HACHE21M009 HACHE21M001 HACHE21M002 HACHB20B300
Ioniselektiivinen elektrodi (ISE), referenssi elektrodit, Radiometer Analytical
Elektrodit ja niiden oheistarvikkeet Ioniselektiiviset elektrodit, ISE
Radiometer Analytical reference electrodes are available with a variety of reference systems and liquid junctions.
  • Liquid junction: porous pin
  • Red-Rod technology provides a reproducible, fast and accurate response
  • Calomel reference electrode can be used for special purposes
* These electrodes contain mercury, please dispose according to regulations.
** Screw cap electrodes require an external connection cable for connection to a meter.