Pipetti, yksikanavainen, säädettävä tilavuus
Toimittaja: Hach
BBP065 BBP078
HACHBBP065EA 620 EUR
HACHBBP065 HACHBBP078
Pipetti, yksikanavainen, säädettävä tilavuus
Pipetit ja pipetintäyttäjät Pipetit
These piston pipettes are handy, general-purpose microliter pipettes for transferring exact volumes of liquid. They operate on the principle of air expulsion, making use of disposable tips. The volume that is to be pipetted is shown in a clearly visible display on the pipette handle.
  • Fitted with mechanical tip ejector system to prevent possible contamination
  • Pipette is operated by means of the push button at the end of the handle
  • Factory calibrated; no recalibration is needed for routine use