Portable water sampler, AS950
Toimittaja: Hach
ASPCXXXC161XX ASPCXXXL111XX ASPCXXXS141XX ASPCXXXC121XX ASPCXXXS171XX
HACHASPCXXXC161XXEA 3370 EUR
HACHASPCXXXC161XX HACHASPCXXXL111XX HACHASPCXXXS141XX HACHASPCXXXC121XX HACHASPCXXXS171XX
Portable water sampler, AS950
Näytteenottimet Vesinäytteenottimet
These lightweight samplers with low power requirements (12 VDC) have been designed for easy mobility.
  • Desiccant and pump tubing easily accessed
  • Large full-colour display for intuitive, single-screen programming, USB upload and download capabilities
  • Copy programs to save time and status screening to eliminate uncertainty
  • Configurable alarms and recorded diagnostics for the measurements (channels): pH, level and power supply
  • Rugged see-through pump cover for rapid inspection
  • FSData Desktop software for data storage
They are suitable for fieldMore Product Information