Filter Paper, Glass Fibre, Extra Thick
Toimittaja: Hach
2842100
HACH2842100EA 127 EUR
HACH2842100
Filter Paper, Glass Fibre, Extra Thick
Suodattimet Suodatinpaperit Kvalitatiivinen suodatinpaperi
For filtration of heavily contaminated samples. High particulate-holding capacity makes discs efficient depth filters and allows for filtration of large volumes of solutions.