Microorganism tests, BART test for acid producing bacteria
Toimittaja: Hach
2831409
HACH2831409EA 201 EUR
HACH2831409
Microorganism tests, BART test for acid producing bacteria
Pikatestit patogeenien ja allergeenien osoittamiseen Mikrobitestit
BART biodetectors are excellent diagnostic tools to help identify the presence and activity of various bacteria.
  • Simple yet effective method for monitoring the population size and/or activity of specific groups of bacteria
  • Easy to use, requiring no elaborate or costly equipment and no specialized training
  • Effective and affordable tests are easy to interpret and can be performed at room temperature in virtually any environment