Tubidity standards for 2100A
Toimittaja: Hach
2659105 2659110
HACH2659105EA 449 EUR
HACH2659105 HACH2659110
Tubidity standards for 2100A
Standardiaineet ja vertailumateriaalit Sameustandardit
Stablcal Stabilised Formazin Turbidity Standards are true formazin dilutions developed for use in any turbidimeter.
  • Precise, pre-mixed formazin solutions
  • Based on Formazin Primary Standards
  • Enhanced stability saves time and labour
  • Confidence in results