Test kits, drop count tests/
Toimittaja: Hach
145201 222301 143601 208601 144001 225401 222702 145701 42632 146900 148002 2291700 2314500 2308100 2408532 2437600 2437800 2440000 2437700 145400 145700 70532 145300 2444300 2687200 2183100 2408549 2437500
HACH145201EA 134 EUR
HACH145201 HACH222301 HACH143601 HACH208601 HACH144001 HACH225401 HACH222702 HACH145701 HACH42632 HACH146900 HACH148002 HACH2291700 HACH2314500 HACH2308100 HACH2408532 HACH2437600 HACH2437800 HACH2440000 HACH2437700 HACH145400 HACH145700 HACH70532 HACH145300 HACH2444300 HACH2687200 HACH2183100 HACH2408549 HACH2437500
Test kits, drop count tests/
Ympäristöanalytiikan pikatestit
The drop count test is a simple, easy and cost-effective way to test for many parameters like chlorine, chloride, dissolved oxygen, carbon dioxide, sulphite, nitrogen, formaldehyde, and more.
  • Portable - ideal for use in field or in lab
  • Used as a quick reference or for confirmatory test with colour indicator
  • Reagents supplied with the kit
Varoitus: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.