Buffer powder pillows
Toimittaja: Hach
2227064 2227066 2227095 2226966 2226995 2227166 2227195 2299264 1410766 1407995 2226964 2227164 1409895
HACH2227064EA 277 EUR
HACH2227064 HACH2227066 HACH2227095 HACH2226966 HACH2226995 HACH2227166 HACH2227195 HACH2299264 HACH1410766 HACH1407995 HACH2226964 HACH2227164 HACH1409895
Buffer powder pillows
Biokemialliset puskuriliuokset pH-puskuriliuokset
The unit-dose powder pillows assure buffer freshness and eliminate the possibility of contamination. Each powder pillow prepares 50 ml of buffer solutions of respective pH.
  • Fresh and reliable
  • No danger of contamination
  • Traceable to NIST standards
Toimitustiedot: The buffer pillows are delivered in an airtight container and are supplied with a certificate of conformity.