pH-puskuriliuokset

Toimittaja: Hach

1222149 1222249 1222349 S11M009 1222220 S11M010 S11M011
HACH1222149EA 22.3 EUR
HACH1222149 HACH1222249 HACH1222349 HACHS11M009 HACH1222220 HACHS11M010 HACHS11M011
pH-puskuriliuokset
Biokemialliset puskuriliuokset pH-puskuriliuokset

For calibration of pH meters.


Accuracy: pH is ±0,02 at 25 °C


Sertifikaatit: Traceable to NIST.


Varoitus: Please note that the pH 4 and 10 buffers contain formaldehyde <0,1% and the pH 7 buffer contains formaldehyde 0,1 to 1%.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR