4-(Trifluoromethyl)piperidine 97%

Toimittaja: Thermo Fisher Scientific
Varoitus

Synonyms: 4-Trifluoromethylpiperidine

H64840.03 H64840.06
H64840.03EA 411 EUR
H64840.03 H64840.06
4-(Trifluoromethyl)piperidine 97%
4-(Trifluoromethyl)piperidine

Purity: 97% CAS No.: 657-36-3 Molecular Formula: C6H10F3N Formula Weight: 153.15 Physical Form: Liquid Appearance: Colourless MDL No.: MFCD00102068

Kaava: C₆H₁₀F₃N
MW: 153.15 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00102068
CAS-numero: 657-36-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR