L-isoleucine ethyl ester hydrochloride 98%

Toimittaja: Thermo Fisher Scientific

H64640.06 H64640.14
H64640.06EA 163 EUR
H64640.06 H64640.14
L-isoleucine ethyl ester hydrochloride 98%
L-isoleucine ethyl ester hydrochloride

Purity: 98% CAS No.: 56782-52-6 Molecular Formula: C8H17NO2.HCl Formula Weight: 195.69 Physical Form: Powder Appearance: White Beilstein Reference: 4710859 MDL No.: MFCD09953729

Kaava: C₈H₁₈ClNO₂
Storage Temperature: Huoneenlämpö
MDL numero: MFCD09953729
CAS-numero: 56782-52-6
EINECS: 260-382-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR