2-[2-(Trifluoromethyl)phenyl]ethylamine 95%

Toimittaja: Thermo Fisher Scientific
Vaara

H64492.03 H64492.06 H64492.14
H64492.03EA 121 EUR
H64492.03 H64492.06 H64492.14
2-[2-(Trifluoromethyl)phenyl]ethylamine 95%
2-[2-(Trifluoromethyl)phenyl]ethylamine

Purity: 95% CAS No.: 774-18-5 Molecular Formula: C9H10F3N Formula Weight: 189.18 Physical Form: Liquid Appearance: Pale yellow Beilstein Reference: 7202211 UN No.: UN2735 MDL No.: MFCD00128232

Kaava: C₉H₁₀F₃N
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00128232
CAS-numero: 774-18-5
EINECS: 212-262-5
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR