2-Bromo-4'-(trifluoromethoxy)acetophenone 97%

Toimittaja: Thermo Fisher Scientific
Varoitus

H64105.03 H64105.06
H64105.03EA 178 EUR
H64105.03 H64105.06
2-Bromo-4'-(trifluoromethoxy)acetophenone 97%
2-Bromo-4'-(trifluoromethoxy)acetophenone

Purity: 97% CAS No.: 103962-10-3 Molecular Formula: C9H6BrF3O2 Formula Weight: 283.04 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Physical Form: Solid Appearance: Pale yellow Melting Point: 48-55° Beilstein Reference: 7479010 MDL No.: MFCD00052333

Kaava: C₉H₆BrF₃O₂
Sulamispiste: 48…55 °C
Leimahduspiste: >110 °C (230 °F)
MDL numero: MFCD00052333
CAS-numero: 103962-10-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR