D-Tyrosine methyl ester hydrochloride 98%

Toimittaja: Thermo Fisher Scientific
Varoitus

H63630.06 H63630.14
H63630.06EA 128 EUR
H63630.06 H63630.14
D-Tyrosine methyl ester hydrochloride 98%
D-Tyrosine methyl ester hydrochloride

Purity: 98% CAS No.: 3728-20-9 Molecular Formula: C10H13NO3.HCl Formula Weight: 231.68 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to off-white MDL No.: MFCD00066124

Kaava: C₁₀H₁₄ClNO₃
MDL numero: MFCD00066124
CAS-numero: 3728-20-9
EINECS: 223-084-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR