2-Bromobenzothiazole 99%

Toimittaja: Thermo Fisher Scientific
Varoitus

H63286.03 H63286.06 H63286.14
H63286.03EA 113 EUR
H63286.03 H63286.06 H63286.14
2-Bromobenzothiazole 99%
2-Bromobenzothiazole

Purity: 99% CAS No.: 2516-40-7 Molecular Formula: C7H4BrNS Formula Weight: 214.08 Flash Point: 110°(230°F) Physical Form: Crystalline solid Appearance: Pale yellow Melting Point: 39-43° MDL No.: MFCD02681887

Kaava: C₇H₄BrNS
Sulamispiste: 39…43 °C
Leimahduspiste: >110 °C (230 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD02681887
CAS-numero: 2516-40-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR