Ethyl 2-Methylquinoline-6-Carboxylate 97%

Toimittaja: Thermo Fisher Scientific

Synonyms: Ethyl 2-methylquinoline-6-carboxylate

H63213.03 H63213.06
H63213.03EA 172 EUR
H63213.03 H63213.06
Ethyl 2-Methylquinoline-6-Carboxylate 97%
Ethyl-2-methylquinoline-6-carboxylate

Purity: 97% CAS No.: 855763-77-8 Molecular Formula: C13H13NO2 Formula Weight: 215.25 Physical Form: Solid MDL No.: MFCD09879697

Kaava: C₁₃H₁₃NO₂
MW: 215.252 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD09879697
CAS-numero: 855763-77-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR