(S)-2-((tert-Butoxycarbonyl)amino)-3-(pyridin-2-yl)propanoic acid 98%

Toimittaja: Thermo Fisher Scientific
Varoitus

H63037.03 H63037.06
H63037.03EA 240 EUR
H63037.03 H63037.06
(S)-2-((tert-Butoxycarbonyl)amino)-3-(pyridin-2-yl)propanoic acid 98%
(S)-2-((tert-Butoxycarbonyl)amino)-3-(pyridin-2-yl)propanoic acid

Purity: 98% CAS No.: 71239-85-5 Molecular Formula: C13H18N2O4 Formula Weight: 266.29 Physical Form: Solid Beilstein Reference: 6416850 Optical Rotation: -16°(c=1 in methanol) MDL No.: MFCD00191190

Kaava: C₁₃H₁₈N₂O₄
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00191190
CAS-numero: 71239-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR