N-Boc-3-(4-pyridyl)-D-alanine 95%

Toimittaja: Thermo Fisher Scientific
Varoitus

H62215.03 H62215.06
H62215.03EA 416 EUR
H62215.03 H62215.06
N-Boc-3-(4-pyridyl)-D-alanine 95%
N-Boc-3-(4-pyridyl)-D-alanine

Purity: 95% CAS No.: 37535-58-3 Molecular Formula: C13H18N2O4 Formula Weight: 266.29 Storage Temperature: 2°C to 8°C Physical Form: Powder Sensitivity: Air and Moisture Sensitive Appearance: White Melting Point: 223-229° Beilstein Reference: 4195478 Optical Rotation: +4.1°(c=0.6 in acetic acid) MDL No.: MFCD00672529

Kaava: C₁₃H₁₈N₂O₄
Sulamispiste: 223…229 °C
MDL numero: MFCD00672529
CAS-numero: 37535-58-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR