Benzyl chloromethyl ether ≥70%, tech.

Toimittaja: Thermo Fisher Scientific
Vaara

H60207.AD H60207.AK
H60207.ADEA 252 EUR
H60207.AD H60207.AK
Benzyl chloromethyl ether ≥70%, tech.
Benzyl chloromethyl ether
Kaava: C₈H₉ClO
Kiehumispiste: 102 °C
Tiheys: 1,126
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00000886
CAS-numero: 3587-60-8
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR