1-Ethyl-3-methylimidazolium acetate 97%

Toimittaja: Thermo Fisher Scientific

H59277.06 H59277.18
H59277.06EA 280 EUR
H59277.06 H59277.18
1-Ethyl-3-methylimidazolium acetate 97%
1-Ethyl-3-methylimidazolium acetate
Kaava: C₈H₁₄N₂O₂
MDL numero: MFCD06798186
CAS-numero: 143314-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR