Phenylmagnesium bromide 3 mol/l

Toimittaja: Thermo Fisher Scientific
Vaara

H54729.AEEA 684 EUR
H54729.AE
Phenylmagnesium bromide 3 mol/l
Phenylmagnesium bromide
Kaava: C₆H₅BrMg
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00000038
CAS-numero: 100-58-3
EINECS: 202-867-2
UN: 2924
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR