1-Chloro-5-(triethylsilyl)-4-pentyne ≥97%

Toimittaja: Thermo Fisher Scientific
Varoitus

H53515.06 H53515.14
H53515.06EA 125 EUR
H53515.06 H53515.14
1-Chloro-5-(triethylsilyl)-4-pentyne ≥97%
1-Chloro-5-(triethylsilyl)-4-pentyne

Purity: 97% CAS No.: 174125-30-5 Molecular Formula: C11H21ClSi Formula Weight: 216.83 MDL No.: MFCD00671352

Kaava: C₁₁H₂₁ClSi
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00671352
CAS-numero: 174125-30-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR