2-Chloro-3-formylpyridine ≥98%

Toimittaja: Thermo Fisher Scientific
Vaara

H53399.03 H53399.06 H53399.14
H53399.03EA 89.9 EUR
H53399.03 H53399.06 H53399.14
2-Chloro-3-formylpyridine ≥98%
2-Chloro-3-formylpyridine

Purity: 98% CAS No.: 36404-88-3 Molecular Formula: C6H4ClNO Formula Weight: 141.56 Flash Point: 110°(230°F) Melting Point: 50-54° MDL No.: MFCD01315308

Kaava: C₆H₄ClNO
Sulamispiste: 50…54 °C
Leimahduspiste: >110 °C (230 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD01315308
CAS-numero: 36404-88-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR