6-Bromobenzothiazole ≥97%

Toimittaja: Thermo Fisher Scientific
Vaara

Synonyms: 6-Bromobenzo[d]thiazole

H52823.MD H52823.03
H52823.MDEA 140 EUR
H52823.MD H52823.03
6-Bromobenzothiazole ≥97%
6-Bromobenzothiazole

Purity: 97% CAS No.: 53218-26-1 Molecular Formula: C7H4BrNS Formula Weight: 214.08 Melting Point: 52-54° MDL No.: MFCD04115372

Kaava: C₇H₄BrNS
MW: 214.09 g/mol
Sulamispiste: 52…54 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD04115372
CAS-numero: 53218-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR