(S)-1-Boc-piperidine-3-carboxylic acid ≥97%

Toimittaja: Thermo Fisher Scientific
Varoitus

H52575.06 H52575.14
H52575.06EA 225 EUR
H52575.06 H52575.14
(S)-1-Boc-piperidine-3-carboxylic acid ≥97%
(S)-1-Boc-piperidine-3-carboxylic acid

Purity: 97% CAS No.: 88495-54-9 Molecular Formula: C11H19NO4 Formula Weight: 229.28 Melting Point: 165-169° Optical Rotation: +57° (c=1 in chloroform)

Kaava: C₁₁H₁₉NO₄
Sulamispiste: 165…169 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD02179172
CAS-numero: 88495-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR