(R)-3-(Aminomethyl)-1-boc-piperidine 97%

Toimittaja: Thermo Fisher Scientific
Varoitus

H52561.MD H52561.03 H52561.06
H52561.MDEA 113 EUR
H52561.MD H52561.03 H52561.06
(R)-3-(Aminomethyl)-1-boc-piperidine 97%
(R)-3-(Aminomethyl)-1-boc-piperidine

Purity: 97% CAS No.: 140645-23-4 Molecular Formula: C11H22N2O2 Formula Weight: 214.31 MDL No.: MFCD03839876

Kaava: C₁₁H₂₂N₂O₂
Storage Temperature: Huoneenlämpö
MDL numero: MFCD03839876
CAS-numero: 140645-23-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR