D-erythro-Sphingosine hydrochloride 97%

Toimittaja: Thermo Fisher Scientific

H52427.MCEA 710 EUR
H52427.MC H52427.ME
D-erythro-Sphingosine hydrochloride 97%
D-erythro-Sphingosine hydrochloride

Purity: 97% CAS No.: 2673-72-5 Molecular Formula: C18H37NO2.HCl Formula Weight: 335.96 MDL No.: MFCD08436977

Kaava: C₁₈H₃₇NO₂·HCl
Storage Temperature: Huoneenlämpö
MDL numero: MFCD08436977
CAS-numero: 2673-72-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR