1-(2-Aminoethyl)-4-methylpiperazine ≥97%

Toimittaja: Thermo Fisher Scientific
Vaara

H52371.MD H52371.03 H52371.06
H52371.MDEA 154 EUR
H52371.MD H52371.03 H52371.06
1-(2-Aminoethyl)-4-methylpiperazine ≥97%
1-(2-Aminoethyl)-4-methylpiperazine

Purity: 97+% CAS No.: 934-98-5 Molecular Formula: C7H17N3 Formula Weight: 143.43 UN No.: 2735 MDL No.: MFCD03701701

Kaava: C₇H₁₇N₃
Storage Temperature: Huoneenlämpö
MDL numero: MFCD03701701
CAS-numero: 934-98-5
EINECS: 213-296-3
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR