Ethyl-2-bromonicotinate 96%

Toimittaja: Thermo Fisher Scientific
Varoitus

H52335.MD H52335.03
H52335.MDEA 217 EUR
H52335.MD H52335.03
Ethyl-2-bromonicotinate 96%
Ethyl-2-bromonicotinate

Purity: 96% CAS No.: 53087-78-8 Molecular Formula: C8H8BrNO2 Formula Weight: 230.07 MDL No.: MFCD04113825

Kaava: C₈H₈BrNO₂
Storage Temperature: Huoneenlämpö
MDL numero: MFCD11506200
CAS-numero: 53087-78-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR