Fmoc-L-3,3-diphenylalanine 95%

Toimittaja: Thermo Fisher Scientific

H52176.03 H52176.06
H52176.03EA 406 EUR
H52176.03 H52176.06
Fmoc-L-3,3-diphenylalanine 95%
Fmoc-L-3,3-diphenylalanine

Purity: 95% CAS No.: 201484-50-6 Molecular Formula: C30H25NO4 Formula Weight: 463.53 Melting Point: 125-129° Beilstein Reference: 8873562 UN No.: UN3077 MDL No.: MFCD00672337

Kaava: C₃₀H₂₅NO₄
Sulamispiste: 125…129 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00672337
CAS-numero: 201484-50-6
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR