Glycine iso-propyl ester hydrochloride (H-Gly-OiPr.HCl) 96%

Toimittaja: Thermo Fisher Scientific

H50249.03 H50249.06
H50249.03EA 80.9 EUR
H50249.03 H50249.06
Glycine iso-propyl ester hydrochloride (H-Gly-OiPr.HCl) 96%
Glycine iso-propyl ester hydrochloride (H-Gly-OiPr.HCl)

Purity: 96% CAS No.: 14019-62-6 Molecular Formula: C5H11NO2.HCl Formula Weight: 153.61 MDL No.: MFCD00151826

Kaava: C₅H₁₂ClNO₂
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00151826
CAS-numero: 14019-62-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR