5-Bromo-2-formylpyridine ≥99%

Toimittaja: Thermo Fisher Scientific
Varoitus

H50154.03 H50154.06
H50154.03EA 172 EUR
H50154.03 H50154.06
5-Bromo-2-formylpyridine ≥99%
5-Bromo-2-formylpyridine

Purity: 99% CAS No.: 31181-90-5 Molecular Formula: C6H4BrNO Formula Weight: 186.01 Melting Point: 94-96° MDL No.: MFCD04112535

Kaava: C₆H₄BrNO
Sulamispiste: 94…96 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD04112535
CAS-numero: 31181-90-5

Order Now

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR