1,3-Di-tert-butyl-2-methoxybenzene 99%

Toimittaja: Thermo Fisher Scientific

H37487.09 H37487.18 H37487.30
H37487.09EA 83.3 EUR
H37487.09 H37487.18 H37487.30
1,3-Di-tert-butyl-2-methoxybenzene 99%
1,3-Di-tert-butyl-2-methoxybenzene

Purity: 99% CAS No.: 1516-95-6 Molecular Formula: C15H24O Formula Weight: 220.35 Physical Form: Liquid Appearance: Colourless to pale yellow Boiling Point: 124°/9mm MDL No.: MFCD20483412

Kaava: C₁₅H₂₄O
Kiehumispiste: 124 °C (9 mmHg)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD20483412
CAS-numero: 1516-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR