3-(m-Tolyl)propionic acid 96%

Toimittaja: Thermo Fisher Scientific
Varoitus

H34395.03 H34395.06
H34395.03EA 136 EUR
H34395.03 H34395.06
3-(m-Tolyl)propionic acid 96%
3-(m-Tolyl)propionic acid

Purity: 96% CAS No.: 3751-48-2 Molecular Formula: C10H12O2 Formula Weight: 164.20 Melting Point: 42-44° MDL No.: MFCD00060326

Kaava: C₁₀H₁₂O₂
Sulamispiste: 42…44 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00060326
CAS-numero: 3751-48-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR