Palladium 5% on carbon Type 87L wetted with ca. 50% water

Toimittaja: Thermo Fisher Scientific
Varoitus

H33434.09 H33434.18
H33434.09EA 392 EUR
H33434.09 H33434.18
Palladium 5% on carbon Type 87L wetted with ca. 50% water
Palladium
Kaava: Pd
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011167
CAS-numero: 7440-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR