2-Metyyli-1-tetraloni 98%

Toimittaja: Thermo Fisher Scientific

H31865.03
H31865.03EA 150 EUR
H31865.03
2-Metyyli-1-tetraloni 98%
2-Metyyli-1-tetraloni

Purity: 98% CAS No.: 1590-08-5 Molecular Formula: C11H12O Formula Weight: 160.22 Flash Point: 110°(230°F) Refractive Index: 1.5520 Boiling Point: 129°/12mm MDL No.: MFCD00001690

Kaava: C₁₁H₁₂O
Kiehumispiste: 129 °C (12 mmHg)
Tiheys: 1,058
Leimahduspiste: >110 °C (230 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00001690
CAS-numero: 1590-08-5
EINECS: 216-461-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR