2-Vinylbenzoic acid 96%

Toimittaja: Thermo Fisher Scientific
Varoitus

H31723.03 H31723.06
H31723.03EA 172 EUR
H31723.03 H31723.06
2-Vinylbenzoic acid 96%
2-Vinylbenzoic acid

Purity: 96% CAS No.: 27326-43-8 Molecular Formula: C9H8O2 Formula Weight: 148.16 Storage Temperature: 2°C to 8°C Melting Point: 94° MDL No.: MFCD02066273

Kaava: C₉H₈O₂
Sulamispiste: 94 °C
MDL numero: MFCD02066273
CAS-numero: 27326-43-8

Order Now

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR