4-Merkapto-4-metyyli-2-pentanoni ≥98%

Toimittaja: Thermo Fisher Scientific
Varoitus

H31693.03 H31693.06 H31693.14
H31693.03EA 103 EUR
H31693.03 H31693.06 H31693.14
4-Merkapto-4-metyyli-2-pentanoni ≥98%
4-Merkapto-4-metyyli-2-pentanoni

Purity: 98% CAS No.: 19872-52-7 Molecular Formula: C6H12SO Formula Weight: 132.23 Storage Temperature: -30°C to -10°C Flash Point: 54°(129°F) Refractive Index: 1.4620 Odour: Unpleasant Boiling Point: 174° UN No.: 3336 MDL No.: MFCD00085208

Kaava: C₆H₁₂OS
Kiehumispiste: 174 °C
Tiheys: 0,961
Leimahduspiste: 54 °C (129 °F)
MDL numero: MFCD00085208
CAS-numero: 19872-52-7
EINECS: 243-386-8
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR