7-Aminoenanthic acid 98%

Toimittaja: Thermo Fisher Scientific

H27519.03 H27519.06
H27519.03EA 283 EUR
H27519.03 H27519.06
7-Aminoenanthic acid 98%
7-Aminoenanthic acid

Purity: 98% CAS No.: 929-17-9 Molecular Formula: H2N(CH2)6CO2H Formula Weight: 145.19 Melting Point: 194-196° Beilstein Reference: 906887

Kaava: C₇H₁₅NO₂
Sulamispiste: 194…196 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008242
CAS-numero: 929-17-9
EINECS: 213-197-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR