4-Methyl-D-phenylalanine 98%

Toimittaja: Thermo Fisher Scientific

H27518.03
H27518.03EA 107 EUR
H27518.03
4-Methyl-D-phenylalanine 98%
4-Methyl-D-phenylalanine

Purity: 98% CAS No.: 49759-61-7 Molecular Formula: C10H13NO2 Formula Weight: 179.22 MDL No.: MFCD00152137

Kaava: C₁₀H₁₃NO₂
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00152137
CAS-numero: 49759-61-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR