4-(Aminomethyl)benzonitrile hydrochloride 97%

Toimittaja: Thermo Fisher Scientific
Vaara

Synonyms: 4-(Aminomethyl)benzonitrile HCl, α-Amino-p-tolunitrile Hydrochloride, 4-(Aminomethyl)benzonitrile Hydrochloride, 4-Cyanobenzylamine Hydrochloride

H27473.14 H27473.06
H27473.14EA 684 EUR
H27473.14 H27473.06
4-(Aminomethyl)benzonitrile hydrochloride 97%
4-(Aminomethyl)benzonitrile hydrochloride

Purity: 97% CAS No.: 15996-76-6 Molecular Formula: C8H8N2.HCl Formula Weight: 168.62 Melting Point: 279-284° MDL No.: MFCD01861472

Kaava: C₈H₉ClN₂
Sulamispiste: 279…284 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD01861472
CAS-numero: 15996-76-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR