2-Bromokinoliini 96%

Toimittaja: Thermo Fisher Scientific
Varoitus

H27302.03 H27302.06 H27302.14
H27302.03EA 177 EUR
H27302.03 H27302.06 H27302.14
2-Bromokinoliini 96%
2-Bromokinoliini

Purity: 96% CAS No.: 2005-43-8 Molecular Formula: C9H6BrN Formula Weight: 208.06 MDL No.: MFCD00234480

Kaava: C₉H₆BrN
MW: 208.06 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00234480
CAS-numero: 2005-43-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR