tert-Butyl-3-oxopyrrolidine-1-carboxylate 97%

Toimittaja: Thermo Fisher Scientific
Varoitus

H27023.09 H27023.03
H27023.09EA 871 EUR
H27023.09 H27023.03
tert-Butyl-3-oxopyrrolidine-1-carboxylate 97%
tert-Butyl-3-oxopyrrolidine-1-carboxylate

Purity: 97% CAS No.: 101385-93-7 Molecular Formula: C9H15NO3 Formula Weight: 185.22 Storage Temperature: 2°C to 8°C Melting Point: 34-38° MDL No.: MFCD01631194

Kaava: C₉H₁₅NO₃
Sulamispiste: 34…38 °C
MDL numero: MFCD01631194
CAS-numero: 101385-93-7

Order Now

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR