2-Fluoroisonicotinonitrile ≥97%

Toimittaja: Thermo Fisher Scientific
Varoitus

Synonyms: 4-Cyano-2-fluoropyridine

H25938.14 H25938.06 H25938.03
H25938.14EA 394 EUR
H25938.14 H25938.06 H25938.03
2-Fluoroisonicotinonitrile ≥97%
2-Fluoroisonicotinonitrile

Purity: 97% CAS No.: 3939-14-8 Molecular Formula: C6H3FN2 Formula Weight: 122.10 Melting Point: 31-33° MDL No.: MFCD09027299

Kaava: C₆H₃FN₂
MW: 122.1 g/mol
Sulamispiste: 31…33 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD09027299
CAS-numero: 3939-14-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR