Acetoxyacetyl chloride ≥97%

Toimittaja: Thermo Fisher Scientific
Vaara

H25797.14 H25797.06 H25797.PG
H25797.14EA 254 EUR
H25797.14 H25797.06 H25797.PG
Acetoxyacetyl chloride ≥97%
Acetoxyacetyl chloride

Purity: 97% CAS No.: 13831-31-7 Molecular Formula: C4H5ClO3 Formula Weight: 136.53 Flash Point: 71°(160°F) Refractive Index: 1.4271 Boiling Point: 55°/12mm UN No.: 3265 MDL No.: MFCD00011535

Kaava: C₄H₅ClO₃
Kiehumispiste: 55 °C (12 mmHg)
Tiheys: 1,270
Leimahduspiste: 71 °C (160 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011535
CAS-numero: 13831-31-7
EINECS: 237-542-4
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR