(4-Nitrophenyl)methanesulphonyl chloride ≥97%

Toimittaja: Thermo Fisher Scientific
Vaara

H25794.06 H25794.03 H25794.14
H25794.06EA 290 EUR
H25794.06 H25794.03 H25794.14
(4-Nitrophenyl)methanesulphonyl chloride ≥97%
(4-Nitrophenyl)methanesulphonyl chloride

Purity: 97% CAS No.: 4025-75-6 Molecular Formula: C7H6ClNO4S Formula Weight: 235.64 Melting Point: 91-92° UN No.: 3261 MDL No.: MFCD00034030

Kaava: C₇H₆ClNO₄S
Sulamispiste: 91…92 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00034030
CAS-numero: 4025-75-6
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR