Vanupuikot, vanupuikot putkella
Toimittaja: Greiner Bio-One
421161_1000 420161_1300 425010_1000
391-3795EA 392 EUR
391-3795 GREI420161_1300 GREI425010_1000
Vanupuikot, vanupuikot putkella
Vanupuikot Näytteenottopuikot putkissa
These swab tubes are suitable for taking non-human bacteriological, serological or cytological samples.
  • Sterilised by irradiation
  • Tubes are made of polystyrene
  • Swabs are made of cotton
Suitable for hygienic controls in food industry as well as in environmental sampling and veterinary research.
Varoitus: *Tällä sivulla olevat tuotteet eivät kuulu EU -direktiivin 2019/904 soveltamisalaan